missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxybenzene-1-sulfonyl chloride, 97%

Product Code. 10614765
Change view
Click to view available options
Amber Glass Bottle; 10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10614765 Amber Glass Bottle; 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10614765 Supplier Thermo Scientific Maybridge Supplier No. BTB04911EA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 23095-05-8
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C7H6BrClO3S
Linear Formula CH3O(Br)C6H3SO2Cl
Quantity 10 g
Synonym 5-bromo-2-methoxybenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 5-bromo-2-methoxy, 5-bromo-2-methoxybenzenesulphonyl chloride, 5-bromo-2-methoxy-benzenesulfonyl chloride, pubchem16355, acmc-1clw8, ksc495e9j, buttpark 100\07-65, 5-bromo-2-methoxyphenyl chlorosulfone, 5-bromo-2methoxybenzenesulfonyl chloride
InChI Key IXSBNNRUQYYMRM-UHFFFAOYSA-N
SMILES COC1=C(C=C(C=C1)Br)S(=O)(=O)Cl
IUPAC Name 5-bromo-2-methoxybenzenesulfonyl chloride
Molecular Weight (g/mol) 285.536
PubChem CID 520020
Formula Weight 285.54
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-methoxybenzene-1-sulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.