missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxybenzenesulfonyl chloride, 98%

Product Code. 11412467
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412467 1 g 1g
11422467 5 g 5g
11432467 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412467 Supplier Thermo Scientific Alfa Aesar Supplier No. A14293.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23095-05-8 | C7H6BrClO3S | 285.536 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23095-05-8
Molecular Formula C7H6BrClO3S
Molecular Weight (g/mol) 285.536
MDL Number MFCD00051768
InChI Key IXSBNNRUQYYMRM-UHFFFAOYSA-N
Synonym 5-bromo-2-methoxybenzene-1-sulfonyl chloride, benzenesulfonyl chloride, 5-bromo-2-methoxy, 5-bromo-2-methoxybenzenesulphonyl chloride, 5-bromo-2-methoxy-benzenesulfonyl chloride, pubchem16355, acmc-1clw8, ksc495e9j, buttpark 100\07-65, 5-bromo-2-methoxyphenyl chlorosulfone, 5-bromo-2methoxybenzenesulfonyl chloride
PubChem CID 520020
IUPAC Name 5-bromo-2-methoxybenzenesulfonyl chloride
SMILES COC1=C(C=C(C=C1)Br)S(=O)(=O)Cl

Specifications

Melting Point 112°C to 115°C
Quantity 1 g
UN Number UN3261
Beilstein 3093014
Formula Weight 285.54
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-methoxybenzenesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.