missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Bromo-2-nitrophenol, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 5-Bromo-2-nitrophenol |
| CAS | 27684-84-0 |
| Molecular Formula | C6 H4 Br N O3 |
| Quantity | 10 g |
| InChI Formula | InChI=1S/C6H4BrNO3/c7-4-1-2-5(8(10)11)6(9)3-4/h1-3,9H |
| SMILES | Oc1cc(Br)ccc1[N+](=O)[O-] |
| IUPAC Name | 5-bromo-2-nitrophenol |
| Molecular Weight (g/mol) | 218 |
| Formula Weight | 216.9375 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction