missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-nitropyridine, 99%, Thermo Scientific Chemicals

Product Code. 10071444 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10071444 5 g Glass bottle 5g
10061444 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10071444 Supplier Thermo Scientific Acros Supplier No. 398640050
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 39856-50-3 | Molecular Formula: C5H3BrN2O2 | Molecular Weight: 203.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39856-50-3
Molecular Formula C5H3BrN2O2
Molecular Weight (g/mol) 203.00
MDL Number MFCD00160411
InChI Key ATXXLNCPVSUCNK-UHFFFAOYSA-N
Synonym 2-nitro-5-bromopyridine, pyridine, 5-bromo-2-nitro, 2-nitro-5-bromo pyridine, 5-bromo-2-nitro-pyridine, 5-brom-2-nitropyridin, pubchem6691, 5-bromo2-nitropyridine, 5-bromo-2-nitropyridin, 3-bromo-6-nitro pyridine, 5-bromo-2-nitro pyridine
PubChem CID 817620
IUPAC Name 5-bromo-2-nitropyridine
SMILES [O-][N+](=O)C1=CC=C(Br)C=N1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 148.0°C to 150.0°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 202.99
Percent Purity 99%
Chemical Name or Material 5-Bromo-2-nitropyridine
compliance-icons
Product Identifier
  • 5-Bromo-2-nitropyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.