missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(tetrahydropyran-4-yloxy)pyridine, 97%, Thermo Scientific™

Product Code. 10143134
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10143134 1 g Amber Glass Bottle 1g
11346815 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10143134 Supplier Thermo Scientific Maybridge Supplier No. CC58310DA
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 494772-07-5
Molecular Formula C10H12BrNO2
Molecular Weight (g/mol) 258.115
MDL Number MFCD09064985
InChI Key ZCTXTBRJCPHMEP-UHFFFAOYSA-N
Synonym 5-bromo-2-tetrahydro-2h-pyran-4-yloxy pyridine, 5-bromo-2-tetrahydro-2h-pyran-4-yl oxy pyridine, 5-bromo-2-tetrahydropyran-4-yloxy pyridine, 5-bromo-2-oxan-4-yloxy pyridine, pyridine,5-bromo-2-tetrahydro-2h-pyran-4-yl oxy, pubchem22061, 5-bromo-2-oxan-4-yl oxy pyridine, 2-tetrahydropyran-4-yloxy-5-bromopridine, 2-tetrahydropyran-4-yloxy-5-bromopyridine, 4-5-bromopyridin-2-yl oxy oxane
PubChem CID 21914391
IUPAC Name 5-bromo-2-(oxan-4-yloxy)pyridine
SMILES C1COCCC1OC2=NC=C(C=C2)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 258.11
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-(tetrahydropyran-4-yloxy)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.