missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-thiophenemethanol, 95%

Product Code. 15428048
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428048 25 g 25g
15418048 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428048 Supplier Thermo Scientific Alfa Aesar Supplier No. H61493.14
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79387-71-6 | C5H5BrOS | 193.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79387-71-6
Molecular Formula C5H5BrOS
Molecular Weight (g/mol) 193.06
MDL Number MFCD06202626
InChI Key DBRSLMCLMMFHEF-UHFFFAOYSA-N
PubChem CID 11435467
IUPAC Name (5-bromothiophen-2-yl)methanol
SMILES OCC1=CC=C(Br)S1

Specifications

Color Yellow
Quantity 25 g
Synonym 5-bromothien-2-yl methanol, 5-bromothiophen-2-yl methanol, 5-bromo-2-thiophenemethanol, 2-thiophenemethanol, 5-bromo, 5-bromothiophene-2-methanol, 5-bromo-2-thienyl methanol, ksc646o4p, 2-thiophenemethanol,5-bromo, 5-bromo-2-thiophenyl methanol, 5-bromo-2-hydroxymethylthiophene
Formula Weight 193.06
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 5-Bromo-2-thiophenemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.