missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-(trifluoromethyl)pyridine, 97%

Product Code. 11349939
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349939 1 g 1g
11359939 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349939 Supplier Thermo Scientific Alfa Aesar Supplier No. B25731.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 436799-32-5 | C6H3BrF3N | 225.996 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 436799-32-5
Molecular Formula C6H3BrF3N
MDL Number MFCD06657686
InChI Key RPFAUCIXZGMCFN-UHFFFAOYSA-N
Synonym 5-bromo-2-trifluoromethyl pyridine, 2-trifluoromethyl-5-bromopyridine, pyridine, 5-bromo-2-trifluoromethyl, trifluoromethyl-5-bromopyridine, trifluoromethyl-5-bromo-2-pyridine, 5-bromo-2-trifluoromethyl-pyridine, 5-bromo-2 trifluoromethyl pyridine, 5-bromo-2-trifluormethyl pyridine, pubchem2164
PubChem CID 2761197
IUPAC Name 5-bromo-2-(trifluoromethyl)pyridine

Specifications

Melting Point 39°C to 40°C
Boiling Point 76°C to 78°C (18 mmHg)
Quantity 1 g
SMILES C1=CC(=NC=C1Br)C(F)(F)F
Molecular Weight (g/mol) 225.996
Formula Weight 226
Percent Purity 97%
Chemical Name or Material 5-Bromo-2-(trifluoromethyl)pyridine
compliance-icons
Product Identifier
  • 5-Bromo-2-trifluoromethylpyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 5-Bromo-2-trifluoromethylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.