missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-3-(chloromethyl)-1-benzothiophene, ≥97%, Thermo Scientific™

Product Code. 10441493
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10441493 250 mg Amber Glass Bottle 250mg
10621285 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10441493 Supplier Thermo Scientific Maybridge Supplier No. CC33524CB
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 852180-53-1
Molecular Formula C9H6BrClS
Molecular Weight (g/mol) 261.561
MDL Number MFCD07772810
InChI Key ZIOALKCKGFEWMX-UHFFFAOYSA-N
PubChem CID 7164583
IUPAC Name 5-bromo-3-(chloromethyl)-1-benzothiophene
SMILES C1=CC2=C(C=C1Br)C(=CS2)CCl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 5-bromo-3-chloromethyl-1-benzothiophene, 5-bromo-3-chloromethyl benzo b thiophene, 5-bromo-3-chloromethyl-benzo b thiophene, benzo b thiophene,5-bromo-3-chloromethyl, benzo b thiophene, 5-bromo-3-chloromethyl, 5-bromo-3-chloromethyl-1-benzo b thiophene
Formula Weight 261.57
Percent Purity 97%
Chemical Name or Material 5-Bromo-3-(chloromethyl)-1-benzothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.