missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4,5,6,7-tetrahydro-2,1,3-benzoxadiazol-4-one, 97%, Thermo Scientific™

Product Code. 10691113
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10691113 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10691113 Supplier Thermo Scientific Maybridge Supplier No. RJC02150CB
 Restricted Item
Estimated Shipment: 15-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 300574-36-1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H5BrN2O2
MDL Number MFCD00168437
Quantity 250 mg
Synonym 5-bromo-4,5,6,7-tetrahydro-2,1,3-benzoxadiazol-4-one, 5-bromo-4-oxo-4,5,6,7-tetrahydrobenzofurazan, 5-bromo-6,7-dihydrobenzo c 1,2,5 oxadiazol-4 5h-one, 2,1,3-benzoxadiazol-4 5h-one,5-bromo-6,7-dihydro, 5-bromo-6,7-dihydro-5h-benzo 1,2,5 oxadiazol-4-one, 2,1,3-benzoxadiazol-4 5h-one, 5-bromo-6,7-dihydro, 5-bromo-6,7-dihydro-2,1,3-benzoxadiazol-4 5h-one #, 5-bromo-5,6,7-trihydrobenzo 1,2-c 1,2,5-oxadiazol-4-one
InChI Key RLBJBTYYLDXIHR-UHFFFAOYNA-N
SMILES BrC1CCC2=NON=C2C1=O
IUPAC Name 5-bromo-4,5,6,7-tetrahydro-2,1,3-benzoxadiazol-4-one
Molecular Weight (g/mol) 217.02
PubChem CID 572916
Formula Weight 217.02
Percent Purity 97%
Chemical Name or Material 5-Bromo-4,5,6,7-tetrahydro-2,1,3-benzoxadiazol-4-one
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.