missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside

Product Code. 15472969
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15472969 250 mg 250mg
15462969 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15472969 Supplier Thermo Scientific Alfa Aesar Supplier No. J60151.MD
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Substrate for beta-galactosidase | CAS: 107021-38-5 | C14H15BrClNO6 | 408.63 g/mol

Substrate for beta-galactosidase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107021-38-5
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.63
MDL Number MFCD00063780
InChI Key OPIFSICVWOWJMJ-UHFFFAOYNA-N
Synonym x-alpha-gal, 5-bromo-4-chloro-3-indolyl-a-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl a-d-galactopyranoside, 5-bromo-4-chloroindoxyl alpha-d-galactoside, 5-bromo-4-chloro-3-indolyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactopyranoside, 2r,3r,4s,5r,6r-2-5-bromo-4-chloro-1h-indol-3-yl oxy-6-hydroxymethyl oxane-3,4,5-triol, x-alpha-d-galactoside, 5-bromo-4-chloro-3-indoxyl-alpha-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl alpha-d-galactoside
PubChem CID 10173103
ChEBI CHEBI:75506
SMILES OCC1OC(OC2=CNC3=CC=C(Br)C(Cl)=C23)C(O)C(O)C1O

Specifications

Color White
Quantity 250 mg
IUPAC Name 2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
Formula Weight 408.63
Physical Form Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-alpha-D-galactoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.