missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside, 98+%

Product Code. 11440127
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
Unit Size:
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440127 50 mg 50mg
11430127 10 mg 10mg
11344669 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440127 Supplier Thermo Scientific Alfa Aesar Supplier No. B21034.MF
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7240-90-6 | C14H15BrClNO6 | 408.629 g/mol

It is a substrate for beta-galactosidase, which cleaves the glycosidic bond to give 5-bromo-4-chloro-3-hydroxy-1H-indole, which immediately dimerises to give an intensely blue product. It is also used to detect activity of this enzyme in histochemistry and bacteriology. It is employed as a colorless, endogenous or exogenous pigment precursors that may be transformed by biological mechanisms into colored compounds. They are used in biochemical assays and in diagnosis as indicators, particularly in the form of enzyme substrates. X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a substrate for beta-galactosidase, which cleaves the glycosidic bond to give 5-bromo-4-chloro-3-hydroxy-1H-indole, which immediately dimerises to give an intensely blue product. It is also used to detect activity of this enzyme in histochemistry and bacteriology. It is employed as a colorless, endogenous or exogenous pigment precursors that may be transformed by biological mechanisms into colored compounds. They are used in biochemical assays and in diagnosis as indicators, particularly in the form of enzyme substrates. X-Gal is the substrate of choice for blue-white selection of recombinant bacterial colonies with the lac+ genotype.

Solubility
Soluble in 100 mM in DMSO.

Notes
Moisture & Light sensitive. Store away from oxidizing agents, heat, water/moisture and light. Store under inert gas. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7240-90-6
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.629
MDL Number MFCD00005666
InChI Key OPIFSICVWOWJMJ-AEOCFKNESA-N
Synonym x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
PubChem CID 65181
ChEBI CHEBI:75055
IUPAC Name (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

Specifications

Melting Point ∼236°C (decomposition)
Quantity 50 mg
Beilstein 1402009
Sensitivity Light and moisture sensitive
Solubility Information Soluble in 100 mM in DMSO.
Optical Rotation −62° (c=1 in 50% DMF)
Formula Weight 408.64
Percent Purity ≥98%
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-beta-D-galactopyranoside

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.