missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99%, pure

Product Code. 10666594 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mg
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
Unit Size:
100mg
10mg
2.5g
500mg
Product Code. Quantity Packaging unitSize
10666594 500 mg Glass bottle 500mg
10256520 10 mg Glass bottle 10mg
10551861 100 mg Glass bottle 100mg
10593694 2.5 g Glass bottle 2.5g
4 options
This item is not returnable. View return policy

Product Code. 10666594

Brand: Thermo Scientific 227775000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7240-90-6 | C14H15BrClNO6 | 408.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7240-90-6
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.63
MDL Number MFCD00005666
InChI Key OPIFSICVWOWJMJ-AEOCFKNESA-N
Synonym x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
PubChem CID 65181
ChEBI CHEBI:75055
IUPAC Name (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 230.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 500 mg
Merck Index 15,10272
Specific Rotation Condition - 64.00 (20.00°C c=1,H2O/DMF (1/1))
Specific Rotation − 64.00
Formula Weight 408.63
Percent Purity 99%
Grade Pure
Physical Form Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-β-D-galactoside
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.