missing translation for 'onlineSavingsMsg'
Learn More

X-Gal, 5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside, Fisher BioReagents™

Product Code. 10234923
Change view
Click to view available options
Quantity:
100 mg
1 g
Packaging:
Glass Bottle
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10234923 100 mg Glass Bottle 100mg
10113253 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10234923 Supplier Fisher BioReagents Supplier No. BP1615100
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For blue/white colony screening, immunoblotting and immunocytochemical assays

  • Chromogenic substrate used for blue-white selection of recombinant bacterial colonies with the lac+ genotype
  • Forms blue precipitate when hydrolyzed by β-galactosidase
  • Soluble in DMF and Methanol
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7240-90-6
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.629
InChI Key OPIFSICVWOWJMJ-AEOCFKNESA-N
Synonym x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
PubChem CID 65181
ChEBI CHEBI:75055
IUPAC Name (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

Specifikationer

Alpha Vector Bromo-4-chloro-3-indolyl-[beta]-D-galactopyranoside (X-Gal)
Melting Point 230°C
Color White
Packaging Glass Bottle
Quantity 100 mg
Merck Index 15, 10272
Specific Rotation -56 to -64° (+ or -)
Formula Weight 408.63
Physical Form Powder or Solid
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl- β-D-galactopyranoside
compliance-icons
Produktbeteckning
  • 5-Bromo-4-chloro-3-indolyl-b-D-galactopyranoside
Signalord
  • Warning
Riskkategorier
  • Acute toxicity Category 4
Faroangivelser
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Skyddsangivelse
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.