missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside, MP Biomedicals™

Product Code. 11496392 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11496392 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11496392 Supplier MP Biomedicals Supplier No. 0215000190
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 7240-90-6
Molecular Formula C14H15BrClNO6
Molecular Weight (g/mol) 408.629
MDL Number MFCD00005666
InChI Key OPIFSICVWOWJMJ-AEOCFKNESA-N
Synonym x-gal, 5-bromo-4-chloro-3-beta-d-galactopyranosyloxy indole, 5-bromo-4-chloro-3-indolyl-beta-d-galactoside, unii-v595og374w, 5-bromo-4-chloro-3-indolyl beta-galactoside, 5-bromo-4-chloroindol-3-yl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactoside, 5-bromo-4-chloro-3-indolyl-beta-d-galactopyranoside, 5-bromo-4-chloro-3-indolyl beta-d-galactopyranoside, indoxyl-gal
PubChem CID 65181
ChEBI CHEBI:75055
IUPAC Name (2S,3R,4S,5R,6R)-2-[(5-bromo-4-chloro-1H-indol-3-yl)oxy]-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES C1=CC(=C(C2=C1NC=C2OC3C(C(C(C(O3)CO)O)O)O)Cl)Br

Specifications

Color White
Assay Percent Range 0.994
Quantity 500 mg
Formula Weight 408.64
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.