missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-4-chloro-3-indolyl phosphate disodium salt, 98%

Product Code. 15470367
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470367 250 mg 250mg
15460367 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15470367 Supplier Thermo Scientific Alfa Aesar Supplier No. J64451.MD
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Substrate for alkaline phosphatase | CAS: 102185-33-1 | C8H4BrClNNa2O4P | 370.43 g/mol

5-Bromo-4-chloro-3-indolyl phosphate disodium salt, is used for the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-4-chloro-3-indolyl phosphate disodium salt, is used for the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry.

Solubility
Soluble in water (50 mg/mL). Insoluble in dimethylformamide.

Notes
Moisture & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102185-33-1
Molecular Formula C8H4BrClNNa2O4P
Molecular Weight (g/mol) 370.43
MDL Number MFCD00036757
InChI Key OAZUOCJOEUNDEK-UHFFFAOYSA-L
PubChem CID 6097197
IUPAC Name disodium;(5-bromo-4-chloro-1H-indol-3-yl) phosphate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=CNC2=CC=C(Br)C(Cl)=C12

Specifications

Color Blue to White
Quantity 250 mg
Sensitivity Light and moisture sensitive
Synonym sodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 5-bromo-4-chloro-3-indolyl phosphate disodium salt, 5-bromo-4-chloro-3-indoxyl phosphate, disodium salt, bcip™, 1h-indol-3-ol, 5-bromo-4-chloro-, 3-dihydrogen phosphate , sodium salt 1:2, disodium 5-bromo-4-chloro-1h-indol-3-yl phosphate, 1h-indol-3-ol,5-bromo-4-chloro-, 3-dihydrogen phosphate , sodium salt 1:2, 5-bromo-4-chloro-3-indolyl phosphate, 5-bromo-4-chloro-3-indolylphosphate disodium salt
Solubility Information Soluble in water (50mg/mL). Insoluble in dimethylformamide.
Formula Weight 370.43
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Bromo-4-chloro-3-indolyl phosphate disodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.