missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-6-methoxynicotinic Acid, TRC

Product Code. 30330999
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330999 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330999 Supplier Toronto Research Chemicals Supplier No. TRCB71109010MG
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Bromo-6-methoxynicotinic Acid
CAS 1186194-46-6
Molecular Formula C7 H6 Br N O3
Quantity 10 mg
InChI Formula InChI=1S/C7H6BrNO3/c1-12-6-5(8)2-4(3-9-6)7(10)11/h2-3H,1H3,(H,10,11)
SMILES COc1ncc(cc1Br)C(=O)O
IUPAC Name 5-bromo-6-methoxypyridine-3-carboxylic acid
Molecular Weight (g/mol) 232.031
Formula Weight 230.953
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.