missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-7-nitroindoline, 98%

Product Code. p-7046970
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11393886

Brand: Thermo Scientific Alfa Aesar L16993.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 80166-90-1 | C8H7BrN2O2 | 243.06 g/mol

5-Bromo-7-nitroindoline is used as pharmaceuticals and intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-7-nitroindoline is used as pharmaceuticals and intermediates.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80166-90-1
Molecular Formula C8H7BrN2O2
Molecular Weight (g/mol) 243.06
MDL Number MFCD00005708
InChI Key VXKXMHDXFLFIFI-UHFFFAOYSA-N
Synonym 5-bromo-7-nitroindoline, 1h-indole, 5-bromo-2,3-dihydro-7-nitro, 5-bromo-7-nitro-1h indoline, 5-bromo-7-nitro-2,3-dihydroindole, 5-bromo-2,3-dihydro-7-nitro-1h-indole, pubchem7507, 5-bromo-7-nitro-indoline
PubChem CID 3018911
IUPAC Name 5-bromo-7-nitro-2,3-dihydro-1H-indole
SMILES [O-][N+](=O)C1=CC(Br)=CC2=C1NCC2

Specifications

Melting Point 128°C to 133°C
Quantity 1 g
UN Number UN2811
Beilstein 1373199
Solubility Information Insoluble in water.
Formula Weight 243.06
Percent Purity 98%
Chemical Name or Material 5-Bromo-7-nitroindoline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.