missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-DL-tryptophan, 99%

Product Code. 10482313 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
500mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10482313 500 mg Glass Bottle 500mg
10725941 1 g Glass Bottle 1g
10686334 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10482313 Supplier Thermo Scientific Acros Supplier No. 107515000
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6548-09-0 | C11H11BrN2O2 | 283.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6548-09-0
Molecular Formula C11H11BrN2O2
Molecular Weight (g/mol) 283.13
MDL Number MFCD00005648
InChI Key KZDNJQUJBMDHJW-UHFFFAOYSA-N
Synonym 5-bromo-dl-tryptophan, 2-amino-3-5-bromo-1h-indol-3-yl propanoic acid, 5-bromotryptophan, tryptophan, 5-bromo, 5-bromo-tryptophan, 2-amino-3-5-bromoindol-3-yl propanoic acid, acmc-20a9ck, 2-amino-3-5-bromo-1h-indol-3-yl-propionic acid, 5-bromo-dl-tryptophan, sr-2-amino-3-5-bromoindolyl propionic acid
PubChem CID 96735
IUPAC Name 2-amino-3-(5-bromo-1H-indol-3-yl)propanoic acid
SMILES NC(CC1=CNC2=CC=C(Br)C=C12)C(O)=O

Specifications

Melting Point 261.0°C to 267.0°C
CAS Min % 98.5
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 500 mg
Formula Weight 283.12
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 5-Bromo-DL-tryptophan
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.