missing translation for 'onlineSavingsMsg'
Learn More

5-Bromobenzo[b]furan-2-carbonyl chloride, 97%, Thermo Scientific™

Product Code. 10219282
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10219282 1 g 1g
10045762 250 mg 250mg
10123724 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10219282 Supplier Thermo Scientific Maybridge Supplier No. CC31202DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 62878-96-0
Molecular Formula C9H4BrClO2
Molecular Weight (g/mol) 259.483
InChI Key FGPAEHRRVKLUHA-UHFFFAOYSA-N
Synonym 5-bromobenzo b furan-2-carbonyl chloride, 5-bromobenzofuran-2-carbonyl chloride
PubChem CID 2795534
IUPAC Name 5-bromo-1-benzofuran-2-carbonyl chloride
SMILES C1=CC2=C(C=C1Br)C=C(O2)C(=O)Cl

Specifications

Assay Percent Range 95%
Quantity 1 g
Formula Weight 259.49
Percent Purity 95%
Chemical Name or Material 5-Bromobenzo[b]furan-2-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.