missing translation for 'onlineSavingsMsg'
Learn More

5-Bromobenzo[b]furan-2-carboxylic acid 97+%, Thermo Scientific™

Product Code. 10188172
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10188172 1 g Amber Glass Bottle 1g
10285524 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10188172 Supplier Thermo Scientific Maybridge Supplier No. CC31201DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 10242-11-2
Molecular Formula C9H5BrO3
Molecular Weight (g/mol) 241.04
InChI Key QKUWZCOVKRUXKX-UHFFFAOYSA-N
Synonym 5-bromobenzofuran-2-carboxylic acid, 2-benzofurancarboxylic acid, 5-bromo, 5-bromobenzo b furan-2-carboxylic acid, 5-bromo-2-carboxy-1-benzofuran, 5-bromo-2-benzofurancarboxylic acid, 2-benzofurancarboxylicacid, 5-bromo, 5-bromo-benzofuran-2-carboxylic acid, acmc-1bxoo, buttpark 34\07-74, 5-bromo benzo furan 2-carboxylic acid
PubChem CID 735247
IUPAC Name 5-bromo-1-benzofuran-2-carboxylic acid
SMILES C1=CC2=C(C=C1Br)C=C(O2)C(=O)O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 241.04
Percent Purity 97%
Chemical Name or Material 5-Bromo-1-benzofuran-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.