missing translation for 'onlineSavingsMsg'
Learn More

5-Bromobenzo[b]thiophene-2-carboxylic acid, ≥97%, Thermo Scientific™

Product Code. 10650695
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10650695 1 g 1g
10127842 250 mg 250mg
10158424 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10650695 Supplier Thermo Scientific Maybridge Supplier No. CC38801DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7312-10-9
Molecular Formula C9H4BrO2S
Molecular Weight (g/mol) 256.09
MDL Number MFCD01929338
InChI Key ONNFNEFYXIPHCA-UHFFFAOYSA-M
Synonym 5-bromo-1-benzothiophene-2-carboxylic acid, 5-bromobenzo b thiophene-2-carboxylic acid, 5-bromo-benzo b thiophene-2-carboxylic acid, benzo b thiophene-2-carboxylic acid, 5-bromo, onnfnefyxiphca-uhfffaoysa-n, 5-bromobenzothiophene-2-carboxylic acid, benzo b thiophene-2-carboxylicacid, 5-bromo, 5-bromo-1-benzo b thiophene-2-carboxylic acid
PubChem CID 737737

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 257.11
Percent Purity 97%
Chemical Name or Material 5-Bromo-1-benzothiophene-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.