missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindole-3-acetic acid, 97%, Thermo Scientific Chemicals

Product Code. 11412270
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11412270 1 g 1g
15453847 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11412270 Supplier Thermo Scientific Alfa Aesar Supplier No. H29217.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40432-84-6
Molecular Formula C10H8BrNO2
Molecular Weight (g/mol) 254.083
MDL Number MFCD00005637
InChI Key WTFGHMZUJMRWBK-UHFFFAOYSA-N
Synonym 5-bromoindole-3-acetic acid, 2-5-bromo-1h-indol-3-yl acetic acid, 5-bromo-1h-indol-3-yl-acetic acid, 1h-indole-3-acetic acid, 5-bromo, 5-bromo-1h-indol-3-yl acetic acid, 5-bromo-3-indoleacetic acid, chembl82440, 5-bromo-1h-indole-3-acetic acid, 5-bromoindole-3-aceticacid, 4ojq
PubChem CID 96734
IUPAC Name 2-(5-bromo-1H-indol-3-yl)acetic acid
SMILES C1=CC2=C(C=C1Br)C(=CN2)CC(=O)O

Specifications

Melting Point 144°C to 145°C (decomposition)
Quantity 1 g
Formula Weight 254.09
Percent Purity 97%
Chemical Name or Material 5-Bromoindole-3-acetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.