missing translation for 'onlineSavingsMsg'
Learn More

5-Bromoindole-3-acetic acid, 99%

Product Code. 10589980 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10589980 250 mg Glass bottle 250mg
10244370 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10589980 Supplier Thermo Scientific Acros Supplier No. 107072500
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 40432-84-6 | C10H8BrNO2 | 254.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40432-84-6
Molecular Formula C10H8BrNO2
Molecular Weight (g/mol) 254.08
MDL Number MFCD00005637
InChI Key WTFGHMZUJMRWBK-UHFFFAOYSA-N
Synonym 5-bromoindole-3-acetic acid, 2-5-bromo-1h-indol-3-yl acetic acid, 5-bromo-1h-indol-3-yl-acetic acid, 1h-indole-3-acetic acid, 5-bromo, 5-bromo-1h-indol-3-yl acetic acid, 5-bromo-3-indoleacetic acid, chembl82440, 5-bromo-1h-indole-3-acetic acid, 5-bromoindole-3-aceticacid, 4ojq
PubChem CID 96734
IUPAC Name 2-(5-bromo-1H-indol-3-yl)acetic acid
SMILES C1=CC2=C(C=C1Br)C(=CN2)CC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 141°C to 148°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 250 mg
Solubility Information Solubility in water: insoluble in water
Formula Weight 254.08
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromoindole-3-acetic acid, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.