missing translation for 'onlineSavingsMsg'
Learn More

5-(Bromomethyl)-2,1,3-benzothiadiazole, 95+%, Thermo Scientific™

Product Code. 10306924
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10306924 10 g 10g
10120872 250 mg 250mg
10292772 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10306924 Supplier Thermo Scientific Maybridge Supplier No. CC09208EA
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 65858-50-6
Molecular Formula C7H5BrN2S
Molecular Weight (g/mol) 229.095
InChI Key JEPACAAYCVWCFI-UHFFFAOYSA-N
PubChem CID 591004
IUPAC Name 5-(bromomethyl)-2,1,3-benzothiadiazole
SMILES C1=CC2=NSN=C2C=C1CBr

Specifications

Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 10 g
Synonym 5-bromomethyl-2,1,3-benzothiadiazole, 5-bromomethyl benzo c 1,2,5 thiadiazole, 5-bromomethylbenzo-2,1,3-thiadiazole, 5-bromomethyl-benzo 1,2,5 thiadiazole, 2,1,3-benzothiadiazole,5-bromomethyl, 2,1,3-benzothiadiazole, 5-bromomethyl, 5-bromomethyl benzo c 1,2,5-thiadiazole, 5-bromomethyl-2,1,3-benzthiadiazole, 5-bromomethylbenzo-2.1.3-thiadiazole, 5-bromomethylbenzo 2,1,3 thiadiazole
Formula Weight 229.1
Percent Purity 95%
Chemical Name or Material 5-(Bromomethyl)-2, 1, 3-benzothiadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.