missing translation for 'onlineSavingsMsg'
Learn More

5-(Bromomethyl)-2,4-diphenyl-1,3-thiazole, Tech., Thermo Scientific™

Product Code. 11350617
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11350617 250 mg 250mg
10664405 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11350617 Supplier Thermo Scientific Maybridge Supplier No. CC42308CB
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 876316-44-8
Molecular Formula C16H12BrNS
Molecular Weight (g/mol) 330.243
MDL Number MFCD08271912
InChI Key QDYJLVQRXQKPKL-UHFFFAOYSA-N
Synonym 5-bromomethyl-2,4-diphenyl-1,3-thiazole, thiazole,5-bromomethyl-2,4-diphenyl, 5-bromomethyl-2,4-diphenylthiazole
PubChem CID 18525758
IUPAC Name 5-(bromomethyl)-2,4-diphenyl-1,3-thiazole
SMILES C1=CC=C(C=C1)C2=C(SC(=N2)C3=CC=CC=C3)CBr

Specifications

CAS Min % 80.0
CAS Max % 100.0
Quantity 250 mg
Formula Weight 330.25
Percent Purity Tech.
Grade Technical
Chemical Name or Material 5-(Bromomethyl)-2, 4-diphenyl-1, 3-thiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.