missing translation for 'onlineSavingsMsg'
Learn More

5-(Bromomethyl)-3-phenylisoxazole, 97%, Thermo Scientific™

Product Code. 10642485
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10642485 250 mg Amber Glass Bottle 250mg
10172934 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10642485 Supplier Thermo Scientific Maybridge Supplier No. CC64508CB
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 2039-50-1
Molecular Formula C10H8BrNO
Molecular Weight (g/mol) 238.084
MDL Number MFCD00159724
InChI Key ANRMBFFXQKJEIS-UHFFFAOYSA-N
PubChem CID 2748796
IUPAC Name 5-(bromomethyl)-3-phenyl-1,2-oxazole
SMILES C1=CC=C(C=C1)C2=NOC(=C2)CBr

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 5-bromomethyl-3-phenylisoxazole, 5-bromomethyl-3-phenyl-1,2-oxazole, isoxazole, 5-bromomethyl-3-phenyl, 5-bromomethyl-3-phenyl-isoxazole, 3-phenyl-5-bromomethylisoxazole, isoxazole,5-bromomethyl-3-phenyl
Formula Weight 238.08
Percent Purity 97%
Chemical Name or Material 5-(Bromomethyl)-3-phenylisoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.