missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopentyl acetate, 98%

Product Code. 11462987
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11462987 100 g 100g
11442987 5 g 5g
11452987 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462987 Supplier Thermo Scientific Alfa Aesar Supplier No. A15549.22
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15848-22-3 | C7H13BrO2 | 209.08 g/mol

5-?Bromopentyl Acetate is used to model the toxicity of haloesters on marine bacterium such as Vibrio fischeri. It is used to produce 5-Bromo-pentan-1-ol, a raw material for chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-​Bromopentyl Acetate is used to model the toxicity of haloesters on marine bacterium such as Vibrio fischeri. It is used to produce 5-Bromo-pentan-1-ol, a raw material for chemical synthesis.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15848-22-3
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00039204
InChI Key JIHJLQWUVZUKCH-UHFFFAOYSA-N
Synonym 5-bromoamyl acetate, 5-bromo-n-amyl acetate, 1-acetoxy-5-bromopentane, 1-pentanol, 5-bromo-,1-acetate, 5-?bromopentyl acetate, acmc-209dhw, 5-bromopentylacetate, 5-bromo-1-pentanyl acetate, 5-bromopentyl acetate, acetic acid 5-bromoamyl ester
PubChem CID 85140
IUPAC Name 5-bromopentyl acetate
SMILES CC(=O)OCCCCCBr

Specifications

Density 1.255
Boiling Point 109°C to 110°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.462
Quantity 100 g
Beilstein 1752109
Solubility Information Slightly soluble in water.
Formula Weight 209.09
Percent Purity 98%
Chemical Name or Material 5-Bromopentyl acetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.