missing translation for 'onlineSavingsMsg'
Learn More

5-Bromophthalide, 98%

Product Code. 15476777
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476777 100 g 100g
15456777 5 g 5g
15466777 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15476777 Supplier Thermo Scientific Alfa Aesar Supplier No. H33891.22
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 64169-34-2 | C8H5BrO2 | 213.03 g/mol

5-Bromophthalide is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is a key intermediate of citalopram.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromophthalide is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is a key intermediate of citalopram.

Solubility
Soluble in acetone.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64169-34-2
Molecular Formula C8H5BrO2
Molecular Weight (g/mol) 213.03
MDL Number MFCD01797360
InChI Key IUSPXLCLQIZFHL-UHFFFAOYSA-N
Synonym 5-bromophthalide, 5-bromoisobenzofuran-1 3h-one, 5-bromo-1 3h-isobenzofuranone, 5-bromo-3h-isobenzofuran-1-one, 1 3h-isobenzofuranone, 5-bromo, 5-bromo-2-benzofuran-1 3h-one, 5-bromo-3-hydroisobenzofuran-1-one, 5-bromo-1,3-dihydro-2-benzofuran-1-one, 5-bromo-1,3-dihydro-isobenzofuran-1-one, 5-bromphthalide
PubChem CID 603144
IUPAC Name 5-bromo-1,3-dihydro-2-benzofuran-1-one
SMILES BrC1=CC=C2C(=O)OCC2=C1

Specifications

Melting Point 162°C to 166°C
Quantity 100 g
Solubility Information Soluble in acetone.
Formula Weight 213.03
Percent Purity 98%
Chemical Name or Material 5-Bromophthalide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.