missing translation for 'onlineSavingsMsg'
Learn More

5-Bromophthalide, 98%

Product Code. 10091592 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10091592 25 g Glass bottle 25g
10332913 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10091592 Supplier Thermo Scientific Acros Supplier No. 373330250
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 64169-34-2 | C8H5BrO2 | 213.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64169-34-2
Molecular Formula C8H5BrO2
Molecular Weight (g/mol) 213.03
MDL Number MFCD01797360
InChI Key IUSPXLCLQIZFHL-UHFFFAOYSA-N
Synonym 5-bromophthalide, 5-bromoisobenzofuran-1 3h-one, 5-bromo-1 3h-isobenzofuranone, 5-bromo-3h-isobenzofuran-1-one, 1 3h-isobenzofuranone, 5-bromo, 5-bromo-2-benzofuran-1 3h-one, 5-bromo-3-hydroisobenzofuran-1-one, 5-bromo-1,3-dihydro-2-benzofuran-1-one, 5-bromo-1,3-dihydro-isobenzofuran-1-one, 5-bromphthalide
PubChem CID 603144
IUPAC Name 5-bromo-3H-2-benzofuran-1-one
SMILES BrC1=CC=C2C(=O)OCC2=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 162°C to 166°C
Color Beige to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 213.03
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromophthalide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.