missing translation for 'onlineSavingsMsg'
Learn More

5-bromopyridine-3-boronic acid, 97%, Maybridge

Product Code. 10346754
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10346754 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10346754 Supplier Thermo Scientific Maybridge Supplier No. MAY00046DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 452972-09-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C5H5BBrNO2
MDL Number MFCD02685634
Quantity 1 g
Synonym 5-bromopyridine-3-boronic acid, 5-bromopyridin-3-yl boronic acid, 3-bromopyridine-5-boronic acid, 3-bromo-5-pyridineboronic acid, 3-bromopyridyl-5-boronic acid, 5-bromo-3-pyridyl boronic acid, 3-bromo-5-pyridylboronic acid, boronic acid, 5-bromo-3-pyridinyl, 5-bromopyridine-3-boronicacid, pubchem5305
InChI Key ICCGFOKNFZWCTJ-UHFFFAOYSA-N
SMILES OB(O)C1=CN=CC(Br)=C1
IUPAC Name (5-bromopyridin-3-yl)boronic acid
Molecular Weight (g/mol) 201.81
PubChem CID 3814811
Formula Weight 201.81
Percent Purity 97%
Chemical Name or Material 5-Bromopyridine-3-boronic acid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.