missing translation for 'onlineSavingsMsg'
Learn More

5-Bromopyridine-3-boronic acid, tech. 85%, Thermo Scientific Chemicals

Product Code. 11354516
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354516 1 g 1g
11374516 250 mg 250mg
11364516 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11354516 Supplier Thermo Scientific Alfa Aesar Supplier No. L20084.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 452972-09-7
Molecular Formula C5H5BBrNO2
Molecular Weight (g/mol) 201.81
MDL Number MFCD02685634
InChI Key ICCGFOKNFZWCTJ-UHFFFAOYSA-N
Synonym 5-bromopyridine-3-boronic acid, 5-bromopyridin-3-yl boronic acid, 3-bromopyridine-5-boronic acid, 3-bromo-5-pyridineboronic acid, 3-bromopyridyl-5-boronic acid, 5-bromo-3-pyridyl boronic acid, 3-bromo-5-pyridylboronic acid, boronic acid, 5-bromo-3-pyridinyl, 5-bromopyridine-3-boronicacid, pubchem5305
PubChem CID 3814811
IUPAC Name (5-bromopyridin-3-yl)boronic acid
SMILES OB(O)C1=CN=CC(Br)=C1

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 9119873
Formula Weight 201.82
Percent Purity 85%
Grade Technical
Chemical Name or Material 5-Bromopyridine-3-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.