missing translation for 'onlineSavingsMsg'
Learn More

5-Bromothiazole, 97%

Product Code. 15451608
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15451608 5 g 5g
15461608 250 mg 250mg
15441608 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15451608 Supplier Thermo Scientific Alfa Aesar Supplier No. H55387.06
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3034-55-7 | C3H2BrNS | 164.02 g/mol

5-Bromothiazole is used as a pharmaceutical intermediate and performance material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromothiazole is used as a pharmaceutical intermediate and performance material.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-55-7
Molecular Formula C3H2BrNS
Molecular Weight (g/mol) 164.02
MDL Number MFCD07787394
InChI Key DWUPYMSVAPQXMS-UHFFFAOYSA-N
PubChem CID 546059
IUPAC Name 5-bromo-1,3-thiazole
SMILES C1=C(SC=N1)Br

Specifications

Density 1.835
Flash Point 76°C (169°F)
Refractive Index 1.5955
Quantity 5 g
Sensitivity Moisture sensitive
Synonym 5-bromothiazole, thiazole, 5-bromo, 5-bromo-thiazole, 5-bromthiazole, thiazole,5-bromo, 5-bromothiazol, 5-bromo thiazole, zlchem 497, pubchem2077, 2-bromo-1,4-thiazole
Solubility Information Soluble in water.
Formula Weight 164.02
Percent Purity 98%
Chemical Name or Material 5-Bromothiazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.