missing translation for 'onlineSavingsMsg'
Learn More

5-Bromothiophene-2-carboxaldehyde, 97%

Product Code. 11435373
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11435373

Brand: Thermo Scientific Alfa Aesar A13381.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4701-17-1 | C5H3BrOS | 191.04 g/mol

5-Bromo-2-thiophenecarboxaldehyde was used to prepare 5-[18F]fluoro-2-2-thiophene carboxaldehyde. Also is used in biological studies as anti-inflammatory and anti-tumor activity of the marine mangrove Rhizophora apiculata.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromo-2-thiophenecarboxaldehyde was used to prepare 5-[18F]fluoro-2-2-thiophene carboxaldehyde. Also is used in biological studies as anti-inflammatory and anti-tumor activity of the marine mangrove Rhizophora apiculata.

Solubility
Soluble in chloroform and methanol.

Notes
Air Sensitive, avoid contact with air/oxygen. Store in cool, dry place in tightly closed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4701-17-1
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.04
MDL Number MFCD00005432
InChI Key GFBVUFQNHLUCPX-UHFFFAOYSA-N
Synonym 5-bromo-2-thiophenecarboxaldehyde, 5-bromothiophene-2-carboxaldehyde, 5-bromo-2-thiophenecarbaldehyde, 5-bromothenaldehyde, 5-bromo-2-formylthiophene, 5-bromo-thiophene-2-carbaldehyde, 2-bromo-5-formylthiophene, 2-thiophenecarboxaldehyde, 5-bromo, 2-bromo-5-thiophenecarboxaldehyde, 5-bromothiophene-2-aldehyde
PubChem CID 78428
IUPAC Name 5-bromothiophene-2-carbaldehyde
SMILES BrC1=CC=C(S1)C=O

Specifications

Density 1.762
Boiling Point 104°C to 106°C (10 mmHg)
Flash Point 98°C (208°F)
Refractive Index 1.639
Quantity 5 g
Beilstein 108404
Sensitivity Air sensitive
Solubility Information Soluble in chloroform and methanol.
Formula Weight 191.05
Percent Purity 97%
Chemical Name or Material 5-Bromothiophene-2-carboxaldehyde
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.