missing translation for 'onlineSavingsMsg'
Learn More

5-Bromothiophene-2-carboxylic acid hydrazide, 97%

Product Code. 15476886
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15476886 25 g 25g
15458548 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476886 Supplier Thermo Scientific Alfa Aesar Supplier No. H61959.14
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 98027-27-1 | C5H5BrN2OS | 221.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98027-27-1
Molecular Formula C5H5BrN2OS
Molecular Weight (g/mol) 221.07
MDL Number MFCD01310798
InChI Key OFXZACLPQYOIAV-UHFFFAOYSA-N
Synonym 5-bromo-2-thiophenecarboxylic acid hydrazide, 5-bromo-thiophene-2-carboxylic acid hydrazide, 5-bromo-thiophene-2-carboxylic acidhydrazide, 5-bromothiophene-2-carboxylic acid hydrazide, 2-thiophenecarboxylic acid, 5-bromo-, hydrazide, pubchem16117, ksc488m6p, 5-bromo-2-thiophenecarboxylic hydrazide, 2-bromothiophene-5-carbohydrazide, 2-thiophenecarboxylicacid, 5-bromo-, hydrazide
PubChem CID 2063863
IUPAC Name 5-bromothiophene-2-carbohydrazide
SMILES NNC(=O)C1=CC=C(Br)S1

Specifications

Quantity 25 g
Formula Weight 221.07
Percent Purity 98%
Chemical Name or Material 5-Bromothiophene-2-carboxylic acid hydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.