missing translation for 'onlineSavingsMsg'
Learn More

5-Bromouridine, 98%, Thermo Scientific Chemicals

Product Code. 11325199
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11325199 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11325199 Supplier Thermo Scientific Alfa Aesar Supplier No. A18507.03
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

5-Bromouridine is often incorporated into RNA in order to detect immunocytochemically and analyzed by cytometry. It possesses anti-viral activity and inhibits the activity of viruses such as human immunodeficiency virus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromouridine is often incorporated into RNA in order to detect immunocytochemically and analyzed by cytometry. It possesses anti-viral activity and inhibits the activity of viruses such as human immunodeficiency virus.

S
TRUSTED_SUSTAINABILITY

Specifications

CAS 957-75-5
Melting Point 191°C to 193°C
Molecular Formula C9H11BrN2O6
MDL Number MFCD00006528
Quantity 1 g
Synonym 5-bromouridine, uridine, 5-bromo, 1-beta-ribofuranosyl-5-bromo-uracil, 5-bromo-1-2 r,3 r,4 s,5 r-3,4-dihydroxy-5-hydroxymethyl tetrahydrofuran-2-yl pyrimidine-2,4 1 h,3 h-dione, bromouridine, 5-bromo-uridine, key8pg1brc, unii-key8pg1brc, 5-bromouracil ribonucleoside, bru
Solubility Information Insoluble aq. soln..
InChI Key AGFIRQJZCNVMCW-UHFFFAOYNA-N
SMILES OCC1OC(C(O)C1O)N1C=C(Br)C(=O)NC1=O
IUPAC Name 5-bromo-1-[(2 R,3 R,4 S,5 R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyrimidine-2,4-dione
Molecular Weight (g/mol) 323.10
PubChem CID 91494
ChEBI CHEBI:20553
Formula Weight 323.1
Percent Purity 98%
Chemical Name or Material 5-Bromouridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.