missing translation for 'onlineSavingsMsg'
Learn More

5-Bromovaleric acid, 97%

Product Code. 11409333
missing translation for 'orderingAttributeHoverText'
Quantity:
10 g
100 g
250 g
missing translation for 'unitSize'
100g
10g
250g
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11409333

missing translation for 'mfr': Thermo Scientific Alfa Aesar A11765.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 2067-33-6 | C5H9BrO2 | 181.029 g/mol

5-Bromovaleric acid is used in the synthesis of inhibitors for aminoglycoside resistant bacteria. 5-Bromovaleric acid is also used in the preparation of sequence specific base pair mimics as topoisomerase IB inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromovaleric acid is used in the synthesis of inhibitors for aminoglycoside resistant bacteria. 5-Bromovaleric acid is also used in the preparation of sequence specific base pair mimics as topoisomerase IB inhibitors.

Solubility
Soluble in ethanol. Insoluble in water.

Notes
Store away from strong bases and oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2067-33-6
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.029
MDL Number MFCD00004414
InChI Key WNXNUPJZWYOKMW-UHFFFAOYSA-N
Synonym 5-bromovaleric acid, pentanoic acid, 5-bromo, 5-bromo-n-valeric acid, valeric acid, 5-bromo, 5-bromovalenc acid, 1-bromo-4-carboxybutane, .delta.-bromovaleric acid, delta-bromovaleric acid, 5-bromo valeric acid, 5-bromoval eric acid
PubChem CID 16368
IUPAC Name 5-bromopentanoic acid
SMILES C(CCBr)CC(=O)O

Specifications

Melting Point 38°C to 41°C
Boiling Point 142°C to 145°C (13 mmHg)
Flash Point >100°C (212°F)
Quantity 100 g
UN Number UN3261
Beilstein 1745664
Solubility Information Soluble in ethanol. Insoluble in water.
Formula Weight 181.03
Percent Purity 97%
Chemical Name or Material 5-Bromovaleric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.