missing translation for 'onlineSavingsMsg'
Learn More

5-Bromovaleric acid, 97%

Product Code. p-3777733 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 10226080

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2067-33-6 | C5H9BrO2 | 181.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2067-33-6
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD00004414
InChI Key WNXNUPJZWYOKMW-UHFFFAOYSA-N
Synonym 5-bromovaleric acid, pentanoic acid, 5-bromo, 5-bromo-n-valeric acid, valeric acid, 5-bromo, 5-bromovalenc acid, 1-bromo-4-carboxybutane, .delta.-bromovaleric acid, delta-bromovaleric acid, 5-bromo valeric acid, 5-bromoval eric acid
PubChem CID 16368
IUPAC Name 5-bromopentanoic acid
SMILES C(CCBr)CC(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 37°C to 41°C
Color Yellow to Brown
Boiling Point 142°C to 145°C (13.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula Br(CH2)4CO2H
Quantity 10 g
Beilstein 02, 303
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol
Formula Weight 181.03
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 5-Bromovaleric acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 5-Bromovaleric acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.