missing translation for 'onlineSavingsMsg'
Learn More

5-Bromovanillin, 98%

Product Code. 11425283
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425283 25 g 25g
11435283 100 g 100g
11445283 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11425283 Supplier Thermo Scientific Alfa Aesar Supplier No. A12861.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2973-76-4 | C8H7BrO3 | 231.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2973-76-4
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.05
MDL Number MFCD00006940
InChI Key KLSHZDPXXKAHIJ-UHFFFAOYSA-N
Synonym 5-bromovanillin, benzaldehyde, 3-bromo-4-hydroxy-5-methoxy, vanillin, 5-bromo, 5-bromo-4-hydroxy-3-methoxybenzaldehyde, 3-bromo-4-hydroxy-5-methoxy-benzaldehyde, unii-508wvs56pw, 3-bromo-4-hydroxy-5-methyloxy benzaldehyde, 5-bromovaniline, 5-bromovanilline
PubChem CID 18099
IUPAC Name 3-bromo-4-hydroxy-5-methoxybenzaldehyde
SMILES COC1=CC(C=O)=CC(Br)=C1O

Specifications

Melting Point 162°C to 166°C
Quantity 25 g
Beilstein 2049761
Sensitivity Air and light sensitive
Formula Weight 231.05
Percent Purity 98%
Chemical Name or Material 5-Bromovanillin
compliance-icons
Product Identifier
  • 5-Bromovanillin
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 5-Bromovanillin

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.