missing translation for 'onlineSavingsMsg'
Learn More

5’-Carboxy Meloxicam, TRC

Product Code. 30134294
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134294 50 mg 50mg
30007286 10 mg 10mg
30011131 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134294 Supplier Toronto Research Chemicals Supplier No. TRCC17872550MG
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130262-93-0
Molecular Formula C14 H11 N3 O6 S2
Molecular Weight (g/mol) 381.38
Synonym 5-Thiazolecarboxylic acid, 2-[[(4-hydroxy-2-methyl-1,1-dioxido-2H-1,2-benzothiazin-3-yl)carbonyl]amino]-, 5-Thiazolecarboxylic acid, 2-[[(4-hydroxy-2-methyl-2H-1,2-benzothiazin-3-yl)carbonyl]amino]-, S,S-dioxide, 2-[[(4-Hydroxy-2-methyl-1,1-dioxido-2H-1,2-benzothiazin-3-yl)carbonyl]amino]-5-thiazolecarboxylic acid, 2H-1,2-Benzothiazine, 5-thiazolecarboxylic acid deriv.
IUPAC Name 2-[(4-hydroxy-2-methyl-1,1-dioxo-1λ^{6},2-benzothiazine-3-carbonyl)amino]-1,3-thiazole-5-carboxylic acid
SMILES CN1C(=C(O)c2ccccc2S1(=O)=O)C(=O)Nc3ncc(s3)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H11N3O6S2/c1-17-10(12(19)16-14-15-6-8(24-14)13(20)21)11(18)7-4-2-3-5-9(7)25(17,22)23/h2-6,18H,1H3,(H,20,21)(H,15,16,19)
Formula Weight 381.0089
Percent Purity >95%
Chemical Name or Material 5'-Carboxy Meloxicam
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.