missing translation for 'onlineSavingsMsg'
Learn More

5-Carboxyfluorescein, 95%

Product Code. 10102864 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10102864 100 mg Glass bottle 100mg
10254864 500 mg Glass bottle 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10102864 Supplier Thermo Scientific Acros Supplier No. 425811000
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
This item is not returnable. View return policy

CAS: 76823-03-5 | C21H12O7 | 376.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76823-03-5
Molecular Formula C21H12O7
Molecular Weight (g/mol) 376.32
MDL Number MFCD00036874
InChI Key NJYVEMPWNAYQQN-UHFFFAOYSA-N
Synonym 5-carboxyfluorescein, 5-fam, 4-carboxyfluorescein, 4 5-carboxyfluorescein, 3',6'-dihydroxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-5-carboxylic acid, 3',6'-dihydroxy-3-oxo-3h-spiro 2-benzofuran-1,9'-xanthene-5-carboxylic acid, 3',6'-dihydroxy-3-oxospiro 2-benzofuran-1,9'-xanthene-5-carboxylic acid, spiro isobenzofuran-1 3h ,9'-9h xanthene-5-carboxylic acid, 3',6-dihydroxy-3-oxo, bidd:gt0758, 5-carboxy fluorescein; 5-fam
PubChem CID 123755
ChEBI CHEBI:51617
IUPAC Name 3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid
SMILES OC(=O)C1=CC=C2C(=C1)C(=O)OC21C2=CC=C(O)C=C2OC2=CC(O)=CC=C12

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Orange
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 100 mg
Formula Weight 376.32
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 5-Carboxyfluorescein
Show More Show Less
compliance-icons
Product Identifier
  • 5-Carboxyfluorescein
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.