missing translation for 'onlineSavingsMsg'
Learn More

5-Carboxyfluorescein-N-hydroxysuccinimide Ester, TRC

Product Code. 30358468
Change view
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30358468 100 mg 100mg
30357016 5 mg -
30352152 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30358468 Supplier Toronto Research Chemicals Supplier No. TRCC178350100MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92557-80-7
Molecular Formula C25 H15 N O9
Molecular Weight (g/mol) 473.39
IUPAC Name (2,5-dioxopyrrolidin-1-yl) 3',6'-dihydroxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylate
SMILES Oc1ccc2c(Oc3cc(O)ccc3C24OC(=O)c5cc(ccc45)C(=O)ON6C(=O)CCC6=O)c1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C25H15NO9/c27-13-2-5-17-19(10-13)33-20-11-14(28)3-6-18(20)25(17)16-4-1-12(9-15(16)24(32)34-25)23(31)35-26-21(29)7-8-22(26)30/h1-6,9-11,27-28H,7-8H2
Formula Weight 473.0747
Chemical Name or Material 5-Carboxyfluorescein-N-hydroxysuccinimide Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.