missing translation for 'onlineSavingsMsg'
Learn More

5-chloro-1,3-dimethyl-1H-pyrazole-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10282882
Change view
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10282882 1 g 1g
10764193 250 mg 250mg
10664395 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10282882 Supplier Thermo Scientific Maybridge Supplier No. MO00061DA
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 27006-82-2
Molecular Formula C6H7ClN2O2
Molecular Weight (g/mol) 174.584
MDL Number MFCD00232043
InChI Key RRWQERXMLIEDKJ-UHFFFAOYSA-N
PubChem CID 2779652
IUPAC Name 5-chloro-1,3-dimethylpyrazole-4-carboxylic acid
SMILES CC1=NN(C(=C1C(=O)O)Cl)C

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 1 g
Synonym 5-chloro-1,3-dimethyl-1h-pyrazole-4-carboxylic acid, 5-chloro-1,3-dimethyl-4-pyrazolecarboxylic acid, pubchem23627, chlorodimethylpyrazolecarboxylicacid, 4-carboxy-5-chloro-1,3-dimethyl-1h-pyrazole, 1h-pyrazole-4-carboxylicacid,5-chloro-1,3-dimethyl, 1h-pyrazole-4-carboxylic acid, 5-chloro-1,3-dimethyl, 1h-pyrazole-4-carboxylicacid, 5-chloro-1,3-dimethyl
Formula Weight 174.59
Percent Purity 97%
Chemical Name or Material 5-Chloro-1, 3-dimethyl-1H-pyrazole-4-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.