missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone, 98%

Product Code. 11450717
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450717 250 g 250g
11430717 10 g 10g
11440717 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11450717 Supplier Thermo Scientific Alfa Aesar Supplier No. B22521.30
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 53786-28-0 | C12H14ClN3O | 251.714 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53786-28-0
Molecular Formula C12H14ClN3O
Molecular Weight (g/mol) 251.714
MDL Number MFCD02684518
InChI Key DOAYWDKFDPSTSV-UHFFFAOYSA-N
Synonym 5-chloro-1-4-piperidyl-2-benzimidazolinone, 5-chloro-1-piperidin-4-yl-1h-benzo d imidazol-2 3h-one, 1-4-piperidinyl-1,3-dihydro-5-chlorobenzimidazolone, 5-chloro-1,3-dihydro-1-4-piperidinyl-2h-benzimidazol-2-one, domperidone impurity a, 5-chloro-1-piperidin-4-yl-1,3-dihydro-2h-benzimidazol-2-one, 5-chloro-1-piperidin-4-yl-1,3-dihydro-benzoimidazol-2-one, 4-5-chloro-2-oxo-2h-benzimidazol-1-yl piperidine, 5-chloro-1-4-piperidyl-3-hydrobenzimidazol-2-one
PubChem CID 104607
IUPAC Name 6-chloro-3-piperidin-4-yl-1H-benzimidazol-2-one
SMILES C1CNCCC1N2C3=C(C=C(C=C3)Cl)NC2=O

Specifications

Melting Point 227°C to 230°C
Quantity 250 g
Formula Weight 251.71
Percent Purity 98%
Chemical Name or Material 5-Chloro-1-(4-piperidinyl)-2-benzimidazolidinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.