missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-methyl-1H-pyrazole-4-carbonyl chloride, ≥95%, Thermo Scientific™

Product Code. 10792103
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10792103 10 g Amber Glass Bottle 10g
10344142 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10792103 Supplier Thermo Scientific Maybridge Supplier No. GK03657EA
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 110763-09-2
Molecular Formula C5H4Cl2N2O
Molecular Weight (g/mol) 179.00
MDL Number MFCD02179947
InChI Key XFDCYGXUXLYMNP-UHFFFAOYSA-N
Synonym 5-chloro-1-methyl-1h-pyrazole-4-carbonyl chloride, 1h-pyrazole-4-carbonylchloride, 5-chloro-1-methyl, acmc-20ea37, 5-chloro-1-methyl-4-pyrazolecarbonyl chloride, 5-chloranyl-1-methyl-pyrazole-4-carbonyl chloride, 1h-pyrazole-4-carbonyl chloride, 5-chloro-1-methyl-9ci
PubChem CID 2735773
IUPAC Name 5-chloro-1-methylpyrazole-4-carbonyl chloride
SMILES CN1N=CC(C(Cl)=O)=C1Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 179.01
Percent Purity 95%
Chemical Name or Material 5-Chloro-1-methyl-1H-pyrazole-4-carbonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.