missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-1-pentyne, 98%

Product Code. 11347865
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11347865 25 g 25g
11337865 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347865 Supplier Thermo Scientific Alfa Aesar Supplier No. L02835.14
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14267-92-6 | C5H7Cl | 102.561 g/mol

5-Chloro-1-pentyne is used with α-diazoesters in a copper-catalyzed synthesis of pyrazoles. It is also used in the synthesis of 5-chloro-2-trimethylstannyl-1-pentene by reacting with Me3SnCu.Me2S.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-1-pentyne is used with α-diazoesters in a copper-catalyzed synthesis of pyrazoles. It is also used in the synthesis of 5-chloro-2-trimethylstannyl-1-pentene by reacting with Me3SnCu.Me2S.

Solubility
Not miscible in water.

Notes
Store at 4°C. Protect from heat. Incompatible with heavy metal powders, heat and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14267-92-6
Molecular Formula C5H7Cl
Molecular Weight (g/mol) 102.561
MDL Number MFCD00001014
InChI Key UXFIKVWAAMKFQE-UHFFFAOYSA-N
Synonym 5-chloro-1-pentyne, 1-pentyne, 5-chloro, 5-chloropentyne, 4-pentynyl chloride, 1-chloro-4-pentine, 1-chloro-4-pentyne, pent-4-ynyl chloride, 5-chloro-pent-1-yne, acmc-1bzfd, pent-4-yn-1-yl chloride
PubChem CID 84308
IUPAC Name 5-chloropent-1-yne
SMILES C#CCCCCl

Specifications

Density 0.968
Boiling Point 113°C
Flash Point 15°C (59°F)
Odor Recognizable
Linear Formula Cl(CH2)3C≡CH
Refractive Index 1.444
Quantity 25 g
UN Number UN1993
Beilstein 1736710
Solubility Information Not miscible in water.
Formula Weight 102.56
Percent Purity 98%
Chemical Name or Material 5-Chloro-1-pentyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.