missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-cyanopyridine, 96%

Product Code. 15432239
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15432239 1 g 1g
15442239 5 g 5g
15452239 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15432239 Supplier Thermo Scientific Alfa Aesar Supplier No. H64846.03
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 89809-64-3 | C6H3ClN2 | 138.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 89809-64-3
Molecular Formula C6H3ClN2
Molecular Weight (g/mol) 138.55
MDL Number MFCD03788835
InChI Key WTHODOKFSYPTKA-UHFFFAOYSA-N
Synonym 5-chloro-2-cyanopyridine, 5-chloropicolinonitrile, 3-chloro-6-cyanopyridine, 2-cyano-5-chloro pyridine, 2-pyridinecarbonitrile, 5-chloro, 2-cyano-5-chloropyridine, 6-cyano-3-chloropyridine, 5-chloro-2-pyridinecarbonitrile, pubchem2107, 5-chloropicolinenitrile
PubChem CID 3833942
IUPAC Name 5-chloropyridine-2-carbonitrile
SMILES ClC1=CN=C(C=C1)C#N

Specifikationer

Melting Point 106°C to 108°C
Quantity 1 g
UN Number UN3439
Beilstein 113764
Formula Weight 138.55
Percent Purity 96%
Chemical Name or Material 5-Chloro-2-cyanopyridine

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.