missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2'-deoxyuridine, MedChemExpress

Product Code. 16420967
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16420967 50 mg 50mg
16405516 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16420967 Supplier Medchem Express Supplier No. HY112669
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 5-Chloro-2'-deoxyuridine, a thymine analog, is to study the potential of hypochlorous acid damage to DNA and DNA precursors.

5-Chloro-2'-deoxyuridine, a thymine analog, is to study the potential of hypochlorous acid damage to DNA and DNA precursors.

Specifications

Chemical Name or Material 5-Chloro-2'-deoxyuridine
CAS 50-90-8
Color White
Purity Grade Notes Research
Molecular Formula C9H11ClN2O5
Quantity 50 mg
Synonym 5-Chlorodeoxyuridine CldU
Solubility Information DMSO : 250 mg/mL (951.84 mM; Need ultrasonic)
SMILES OC[C@@H]1[C@H](C[C@H](N2C(NC(C(Cl)=C2)=O)=O)O1)O
Molecular Weight (g/mol) 262.65
Formula Weight 262.65
Percent Purity 99.18%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.