missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxy-4-methylbenzenesulfonyl chloride, Thermo Scientific™

Product Code. 10027719
Change view
Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10027719 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10027719 Supplier Thermo Scientific Maybridge Supplier No. MAY00163DE
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 889939-48-4
Packaging Amber Glass Bottle
Molecular Formula C8H8Cl2O3S
Quantity 5 g
Synonym 5-chloro-2-methoxy-4-methylbenzenesulfonylchloride, 5-chloro-2-methoxy-4-methylbenzene-1-sulfonyl chloride, pubchem5095, acmc-20am4m, 5-chloro-2-methoxy-4-methylphenylsulfonyl chloride, chloro 5-chloro-2-methoxy-4-methylphenyl sulfone, 5-chloranyl-2-methoxy-4-methyl-benzenesulfonyl chloride
InChI Key BJRJHJZVSWZXIP-UHFFFAOYSA-N
SMILES CC1=CC(=C(C=C1Cl)S(=O)(=O)Cl)OC
IUPAC Name 5-chloro-2-methoxy-4-methylbenzenesulfonyl chloride
Molecular Weight (g/mol) 255.109
PubChem CID 17389862
Formula Weight 255.12
Percent Purity 97%
Chemical Name or Material 5-Chloro-2-methoxy-4-methylbenzenesulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.