missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxybenzenamine, TRC

Product Code. 30361615
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
1g
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361615 5 g -
30368094 1 g 1g
30359830 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30361615 Supplier Toronto Research Chemicals Supplier No. TRCC9503405G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-03-4
Molecular Formula C7 H8 Cl N O
Molecular Weight (g/mol) 157.6
Synonym o-Anisidine, 5-chloro- (7CI,8CI), 5-Chloro-2-methoxybenzenamine, (5-Chloro-2-methoxyphenyl)amine, 1-Amino-5-chloro-2-methoxybenzene, 2-Amino-4-chloro-1-methoxybenzene, 2-Amino-4-chloroanisole, 2-Methoxy-5-chloroaniline, 4-Chloro-2-aminoanisole, 5-Chloro-2-methoxyaniline, 5-Chloro-o-anisidine, C.I. 37120, free base, NSC 6114, Tulabase Fast Red R, Tulabase Fast Red RC
IUPAC Name 5-chloro-2-methoxyaniline
SMILES COc1ccc(Cl)cc1N

Specifications

Quantity 5 g
InChI Formula InChI=1S/C7H8ClNO/c1-10-7-3-2-5(8)4-6(7)9/h2-4H,9H2,1H3
Formula Weight 157.0294
Chemical Name or Material 5-Chloro-2-methoxyaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.