missing translation for 'onlineSavingsMsg'
Learn More

5-Chloro-2-methoxyphenylmagnesium bromide, 0.50M in 2-MeTHF

Product Code. 15421266
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15421266 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15421266 Supplier Thermo Scientific Alfa Aesar Supplier No. H54501.AE
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 419535-75-4 | C7H6BrClMgO | 245.78 g/mol

5-Chloro-2-methoxyphenylmagnesium bromide is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloro-2-methoxyphenylmagnesium bromide is used as an organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents
TRUSTED_SUSTAINABILITY

Specifications

CAS 419535-75-4
Molecular Formula C7H6BrClMgO
MDL Number MFCD11113495
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Synonym 5-chloro-2-methoxyphenylmagnesium bromide, gafvlsuafzzumw-uhfffaoysa-m, bromo 5-chloro-2-methoxyphenyl magnesium, 5-chloro-2-methoxyphenylmagnesium bromide, 0.50m in 2-methf, grignard reagent
Solubility Information It reacts with water.
InChI Key GAFVLSUAFZZUMW-UHFFFAOYSA-M
SMILES COC1=CC=C(Cl)C=C1[Mg]Br
IUPAC Name magnesium;1-chloro-4-methoxybenzene-5-ide;bromide
Molecular Weight (g/mol) 245.78
PubChem CID 11207325
Formula Weight 245.79
Concentration or Composition (by Analyte or Components) 0.50M in 2-MeTHF
Chemical Name or Material 5-Chloro-2-methoxyphenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.